missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenoic Acid 98.0+%, TCI America™

Catalog No. p-7130775
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08755G 5 g
H087525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08755G Supplier TCI America Supplier No. H08755G

Chemical Identifiers

CAS 1577-22-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00046558
InChI Key XUDOZULIAWNMIU-UHFFFAOYSA-N
Synonym 5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8
PubChem CID 15308
IUPAC Name hex-5-enoic acid
SMILES C=CCCCC(=O)O

Specifications

Color Colorless
Boiling Point 203°C
Quantity 5 g
UN Number 3265
Formula Weight 114.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Hexenoic Acid
EINECSNumber (2)-0609
TSCA No