Learn More
5-Hexenoic acid, 99%
CAS: 1577-22-6 | C6H10O2 | 114.144 g/mol
$103.75 - $339.49
Chemical Identifiers
| CAS | 1577-22-6 |
|---|---|
| Molecular Formula | C6H10O2 |
| Molecular Weight (g/mol) | 114.144 |
| MDL Number | MFCD00046558 |
| InChI Key | XUDOZULIAWNMIU-UHFFFAOYSA-N |
| Synonym | 5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8 |
| PubChem CID | 15308 |
| IUPAC Name | hex-5-enoic acid |
| SMILES | C=CCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAL1286303
|
N/A | 1 g |
Each for $103.75
|
|
|||||
|
AAL1286306
|
N/A | 5 g |
Each for $339.49
|
|
|||||
Description
5-Hexenoic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Hexenoic acid is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| C6H10O2 | |
| MFCD00046558 | |
| 5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8 | |
| hex-5-enoic acid |
Specifications
| 1577-22-6 | |
| 107°C (17 mmHg) | |
| C6H10O2 | |
| MFCD00046558 | |
| 1743172 | |
| Insoluble in water. | |
| C=CCCCC(=O)O | |
| 114.144 | |
| 114.14 | |
| 5-Hexenoic acid |
| 0.961 | |
| 104°C (219°F) | |
| 1.434 | |
| UN3265 | |
| 5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8 | |
| XUDOZULIAWNMIU-UHFFFAOYSA-N | |
| hex-5-enoic acid | |
| 15308 | |
| 99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
TSCA : No
RUO – Research Use Only