missing translation for 'onlineSavingsMsg'
Learn More

5-Hexenoic acid, 99%

Catalog No. AAL1286303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1286303 1 g
AAL1286306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1286303 Supplier Thermo Scientific Chemicals Supplier No. L1286303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1577-22-6 | C6H10O2 | 114.144 g/mol

5-Hexenoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hexenoic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1577-22-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00046558
InChI Key XUDOZULIAWNMIU-UHFFFAOYSA-N
Synonym 5-hexenoic acid, delta-hexenoic acid, acmc-1bnq8
PubChem CID 15308
IUPAC Name hex-5-enoic acid
SMILES C=CCCCC(=O)O

Specifications

Density 0.961
Boiling Point 107°C (17 mmHg)
Flash Point 104°C (219°F)
Refractive Index 1.434
Quantity 1 g
UN Number UN3265
Beilstein 1743172
Solubility Information Insoluble in water.
Formula Weight 114.14
Percent Purity 99%
Chemical Name or Material 5-Hexenoic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No

RUO – Research Use Only