missing translation for 'onlineSavingsMsg'
Learn More

5-Formylsalicylic Acid 98.0+%, TCI America™

Catalog No. p-7129663
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04005G 5 g
F040025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04005G Supplier TCI America Supplier No. F04005G
Only null left

Chemical Identifiers

CAS 616-76-2
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00006945
InChI Key UTCFOFWMEPQCSR-UHFFFAOYSA-N
Synonym 5-formylsalicylic acid, 2-hydroxy-5-formylbenzoic acid, benzoic acid, 5-formyl-2-hydroxy, 5-formylsalicylicacid, 5-formylsalicylic acide, 3-carboxy-4-hydroxybenzaldehyde, 5-formyl-2-hydroxy-benzoic acid, pubchem10569, acmc-209mva, 5-formyl salicylic acid
PubChem CID 69226
IUPAC Name 5-formyl-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(C=O)=CC=C1O

Specifications

Melting Point 250°C
Color Yellow
Quantity 5 g
Formula Weight 166.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Formylsalicylic Acid
EINECSNumber (3)-1669
TSCA No