accessibility menu, dialog, popup

UserName

HOOOHO

CAS RN 616-76-2

IUPAC Name: 5-formyl-2-hydroxybenzoic acid
Synonyms: 5-formyl-2-hydroxybenzoic acid
Molecular Weight (g/mol): 166.13
Molecular Formula: C8H6O4
InChi Key: UTCFOFWMEPQCSR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(C=O)=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results