Learn More
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%
CAS: 253870-02-9 | C8H9NO3 | 167.164 g/mol
Supplier: Thermo Scientific Chemicals H6371106
Description
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
| 5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid | |
| C8H9NO3 | |
| 5 g | |
| Slightly soluble in water. | |
| CC1=C(NC(=C1C(=O)O)C)C=O | |
| 167.164 | |
| 167.16 | |
| Solid |
| 253870-02-9 | |
| MFCD06202342 | |
| 5-formyl-2,4-dimethylpyrrole-3-carboxylic acid, 2,4-dimethyl-5-formylpyrrole-3-carboxylic acid, 3,5-dimethyl-2-formylpyrrole-4-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl, 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylicacid, 3,5-dimethyl-2-formyl-1h-pyrrole-4-carboxylic acid, 3,5-dimethyl-2-formylindole-4-carboxylic acid, zlchem 714, pubchem9414, acmc-209zxq | |
| YCIHQDVIAISDPS-UHFFFAOYSA-N | |
| 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid | |
| 11073792 | |
| 96% |
Chemical Identifiers
| 253870-02-9 | |
| 167.164 | |
| YCIHQDVIAISDPS-UHFFFAOYSA-N | |
| 11073792 | |
| CC1=C(NC(=C1C(=O)O)C)C=O |
| C8H9NO3 | |
| MFCD06202342 | |
| 5-formyl-2,4-dimethylpyrrole-3-carboxylic acid, 2,4-dimethyl-5-formylpyrrole-3-carboxylic acid, 3,5-dimethyl-2-formylpyrrole-4-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl, 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylicacid, 3,5-dimethyl-2-formyl-1h-pyrrole-4-carboxylic acid, 3,5-dimethyl-2-formylindole-4-carboxylic acid, zlchem 714, pubchem9414, acmc-209zxq | |
| 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only