missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%

Catalog No. p-7055191
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63711MD 250 mg
AAH6371103 1 g
AAH6371106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH63711MD Supplier Thermo Scientific Chemicals Supplier No. H63711MD
Only null left

CAS: 253870-02-9 | C8H9NO3 | 167.164 g/mol

5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 253870-02-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD06202342
InChI Key YCIHQDVIAISDPS-UHFFFAOYSA-N
Synonym 5-formyl-2,4-dimethylpyrrole-3-carboxylic acid, 2,4-dimethyl-5-formylpyrrole-3-carboxylic acid, 3,5-dimethyl-2-formylpyrrole-4-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl, 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylicacid, 3,5-dimethyl-2-formyl-1h-pyrrole-4-carboxylic acid, 3,5-dimethyl-2-formylindole-4-carboxylic acid, zlchem 714, pubchem9414, acmc-209zxq
PubChem CID 11073792
IUPAC Name 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
SMILES CC1=C(NC(=C1C(=O)O)C)C=O

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 167.16
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only