accessibility menu, dialog, popup

UserName

CAS RN 253870-02-9

IUPAC Name: 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
Synonyms: 5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: YCIHQDVIAISDPS-UHFFFAOYSA-N
SMILES: CC1=C(C(O)=O)C(C)=C(N1)C=O
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)