missing translation for 'onlineSavingsMsg'
Learn More

5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid, 96%

CAS: 253870-02-9 | C8H9NO3 | 167.164 g/mol

Supplier:  Thermo Scientific Chemicals H6371103

 View more versions of this product

Catalog No. AAH6371103

Only null left
Explore available promotions

Description

Description

5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid employed as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Specifications

5-Formyl-2,4-dimethylpyrrole-3-carboxylic acid
C8H9NO3
1 g
Slightly soluble in water.
CC1=C(NC(=C1C(=O)O)C)C=O
167.164
167.16
Solid
253870-02-9
MFCD06202342
5-formyl-2,4-dimethylpyrrole-3-carboxylic acid, 2,4-dimethyl-5-formylpyrrole-3-carboxylic acid, 3,5-dimethyl-2-formylpyrrole-4-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl, 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylicacid, 3,5-dimethyl-2-formyl-1h-pyrrole-4-carboxylic acid, 3,5-dimethyl-2-formylindole-4-carboxylic acid, zlchem 714, pubchem9414, acmc-209zxq
YCIHQDVIAISDPS-UHFFFAOYSA-N
5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
11073792
96%

Chemical Identifiers

253870-02-9
167.164
YCIHQDVIAISDPS-UHFFFAOYSA-N
11073792
CC1=C(NC(=C1C(=O)O)C)C=O
C8H9NO3
MFCD06202342
5-formyl-2,4-dimethylpyrrole-3-carboxylic acid, 2,4-dimethyl-5-formylpyrrole-3-carboxylic acid, 3,5-dimethyl-2-formylpyrrole-4-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 5-formyl-2,4-dimethyl, 5-formyl-2,4-dimethyl-1h-pyrrole-3-carboxylicacid, 3,5-dimethyl-2-formyl-1h-pyrrole-4-carboxylic acid, 3,5-dimethyl-2-formylindole-4-carboxylic acid, zlchem 714, pubchem9414, acmc-209zxq
5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only