Learn More
5-(Fmoc-amino)valeric acid, 98%
CAS: 123622-48-0 | C20H21NO4 | 339.39 g/mol
Supplier: Thermo Scientific Chemicals H6626406
Description
5-(Fmoc-amino)valeric acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-(Fmoc-amino)valeric acid is used as pharmaceutical intermediate.
Notes
Protect from heat. Store away from oxidizing agents. Refrigerate. Incompatible with
Specifications
| 5-(Fmoc-amino)valeric acid | |
| White | |
| MFCD00235889 | |
| 3565625 | |
| ULLSWWGYZWBPHK-UHFFFAOYSA-N | |
| 5-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid | |
| 2756098 | |
| 98% |
| 123622-48-0 | |
| C20H21NO4 | |
| 5 g | |
| fmoc-5-ava-oh, 5-9h-fluoren-9-yl methoxy carbonyl amino pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino, ambotzfaa1651, n-fmoc-5-aminopentanoic acid, 5-fmoc-amino pentanoic acid, 5-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid, fmoc-5-ava-oh fmoc-5-aminopentanoic acid | |
| OC(=O)CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 339.39 | |
| 339.39 | |
| Powder |
Chemical Identifiers
| 123622-48-0 | |
| 339.39 | |
| ULLSWWGYZWBPHK-UHFFFAOYSA-N | |
| 2756098 | |
| OC(=O)CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| C20H21NO4 | |
| MFCD00235889 | |
| fmoc-5-ava-oh, 5-9h-fluoren-9-yl methoxy carbonyl amino pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, pentanoic acid, 5-9h-fluoren-9-ylmethoxy carbonyl amino, ambotzfaa1651, n-fmoc-5-aminopentanoic acid, 5-fmoc-amino pentanoic acid, 5-9h-fluoren-9-ylmethoxycarbonylamino pentanoic acid, fmoc-5-ava-oh fmoc-5-aminopentanoic acid | |
| 5-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only