accessibility menu, dialog, popup

UserName

OOHNOOH

CAS RN 123622-48-0

IUPAC Name: 5-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoic acid
Synonyms: 5-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}pentanoic acid
Molecular Weight (g/mol): 339.39
Molecular Formula: C20H21NO4
InChi Key: ULLSWWGYZWBPHK-UHFFFAOYSA-N
SMILES: OC(=O)CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)