missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluorouridine 98.0+%, TCI America™
$68.34 - $218.34
Chemical Identifiers
| CAS | 316-46-1 |
|---|---|
| Molecular Formula | C9H11FN2O6 |
| Molecular Weight (g/mol) | 262.193 |
| MDL Number | MFCD00036832 |
| InChI Key | FHIDNBAQOFJWCA-UAKXSSHOSA-N |
| Synonym | 5-fluorouridine, 5-fur, uridine, 5-fluoro, furd, 5-fluoro-uridine, 5-fluorouracil 1beta-d-ribofuranoside, fluorouridine, chembl54918, 5-fluoro-1-beta-d-ribofuranosyl uracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-fluoropyrimidine-2,4 1h,3h-dione |
| PubChem CID | 9427 |
| ChEBI | CHEBI:40154 |
| IUPAC Name | 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoropyrimidine-2,4-dione |
| SMILES | C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)F |
Chemical Identifiers
| 316-46-1 | |
| 262.193 | |
| FHIDNBAQOFJWCA-UAKXSSHOSA-N | |
| 9427 | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoropyrimidine-2,4-dione |
| C9H11FN2O6 | |
| MFCD00036832 | |
| 5-fluorouridine, 5-fur, uridine, 5-fluoro, furd, 5-fluoro-uridine, 5-fluorouracil 1beta-d-ribofuranoside, fluorouridine, chembl54918, 5-fluoro-1-beta-d-ribofuranosyl uracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-fluoropyrimidine-2,4 1h,3h-dione | |
| CHEBI:40154 | |
| C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)F |
Specifications
| 316-46-1 | |
| White-Yellow | |
| MFCD00036832 | |
| 5-fluorouridine, 5-fur, uridine, 5-fluoro, furd, 5-fluoro-uridine, 5-fluorouracil 1beta-d-ribofuranoside, fluorouridine, chembl54918, 5-fluoro-1-beta-d-ribofuranosyl uracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-fluoropyrimidine-2,4 1h,3h-dione | |
| C1=C(C(=O)NC(=O)N1C2C(C(C(O2)CO)O)O)F | |
| 262.193 | |
| CHEBI:40154 | |
| ≥98.0% (T) | |
| 5-Fluorouridine |
| 185°C | |
| C9H11FN2O6 | |
| 1 g | |
| FHIDNBAQOFJWCA-UAKXSSHOSA-N | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-fluoropyrimidine-2,4-dione | |
| 9427 | |
| 262.19 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : YU8050000
TSCA : No