missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoro-8-quinolinol 99.0+%, TCI America™
Supplier: TCI America F00385G
Specifications
| 5-Fluoro-8-quinolinol | |
| 110°C | |
| C9H6FNO | |
| 5 g | |
| YHXLEKUJMPEQAJ-UHFFFAOYSA-N | |
| 5-fluoroquinolin-8-ol | |
| 217907 | |
| ≥99.0% (GC,T) |
| 387-97-3 | |
| White-Yellow | |
| MFCD00059754 | |
| 5-Fluorooxine | |
| C1=CC2=C(C=CC(=C2N=C1)O)F | |
| 163.151 | |
| 163.15 | |
| Crystalline Powder |
Chemical Identifiers
| 387-97-3 | |
| 163.151 | |
| YHXLEKUJMPEQAJ-UHFFFAOYSA-N | |
| 217907 | |
| C1=CC2=C(C=CC(=C2N=C1)O)F |
| C9H6FNO | |
| MFCD00059754 | |
| 5-Fluorooxine | |
| 5-fluoroquinolin-8-ol |
Safety and Handling
TSCA : No