accessibility menu, dialog, popup

UserName

OHFN

CAS RN 387-97-3

IUPAC Name: 5-fluoroquinolin-8-ol
Synonyms: 8QUINOLINOL5fluoro
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H6FNO
InChi Key: YHXLEKUJMPEQAJ-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results