missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-8-quinolinol 99.0+%, TCI America™

Catalog No. p-7129454
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00381G 1 g
F00385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00381G Supplier TCI America Supplier No. F00381G
Only null left

Chemical Identifiers

CAS 387-97-3
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.151
MDL Number MFCD00059754
InChI Key YHXLEKUJMPEQAJ-UHFFFAOYSA-N
Synonym 5-Fluorooxine
PubChem CID 217907
IUPAC Name 5-fluoroquinolin-8-ol
SMILES C1=CC2=C(C=CC(=C2N=C1)O)F

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 1 g
Formula Weight 163.15
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-8-quinolinol