missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7129835
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06945G 5 g
F069425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06945G Supplier TCI America Supplier No. F06945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 320-98-9
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD00055635
InChI Key GHYZIXDKAPMFCS-UHFFFAOYSA-N
Synonym 5-fluoro-2-nitrobenzoic acid, 2-nitro-5-fluorobenzoic acid, 3-fluoro-6-nitrobenzoic acid, 5-fluoro-2-nitro-benzoic acid, benzoic acid, 5-fluoro-2-nitro, 2-carboxy-4-fluoronitrobenzene, pubchem1309, acmc-1aiv3, 5-fluoro-2nitrobenzoic acid, 5-fluor-2-nitrobenzoic acid
PubChem CID 2737420
SMILES C1=CC(=C(C=C1F)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 136°C
Quantity 5 g
Formula Weight 185.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-nitrobenzoic Acid