accessibility menu, dialog, popup

UserName

CAS RN 320-98-9

IUPAC Name: 5-fluoro-2-nitrobenzoate
Synonyms: 5-fluoro-2-nitrobenzoate
Molecular Weight (g/mol): 184.1
Molecular Formula: C7H3FNO4
InChi Key: GHYZIXDKAPMFCS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=CC=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)