missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methylbenzaldehyde 95.0+%, TCI America™

Catalog No. F07465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07465G 5 g
F074625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07465G Supplier TCI America Supplier No. F07465G
Only null left

Chemical Identifiers

CAS 22062-53-9
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.141
MDL Number MFCD02261763
InChI Key MBOXPKNOGZJXPK-UHFFFAOYSA-N
Synonym 5-fluoro-2-methyl-benzaldehyde, benzaldehyde, 5-fluoro-2-methyl, zlchem 996, pubchem1458, acmc-209fri, ksc494k1f, 2-methyl-5-fluorobenzaldehyde, 5-fluoro-2-methylbenzaldehyde
PubChem CID 2759012
IUPAC Name 5-fluoro-2-methylbenzaldehyde
SMILES CC1=C(C=C(C=C1)F)C=O

Specifications

Color Yellow
Boiling Point 99°C
Quantity 5 g
Formula Weight 138.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-2-methylbenzaldehyde