CAS RN 22062-53-9
IUPAC Name:
5-fluoro-2-methylbenzaldehyde
Synonyms:
5-fluoro-2-methylbenzaldehyde
Molecular Weight (g/mol):
138.14
Molecular Formula:
C8H7FO
InChi Key:
MBOXPKNOGZJXPK-UHFFFAOYSA-N
SMILES:
CC1=CC=C(F)C=C1C=O
1
–
1
of
1
results
1
5-Fluoro-2-methylbenzaldehyde 95.0+%, TCI America™
CAS: 22062-53-9 Molecular Formula: C8H7FO Molecular Weight (g/mol): 138.141 MDL Number: MFCD02261763 InChI Key: MBOXPKNOGZJXPK-UHFFFAOYSA-N Synonym: 5-fluoro-2-methyl-benzaldehyde,benzaldehyde, 5-fluoro-2-methyl,zlchem 996,pubchem1458,acmc-209fri,ksc494k1f,2-methyl-5-fluorobenzaldehyde,5-fluoro-2-methylbenzaldehyde PubChem CID: 2759012 IUPAC Name: 5-fluoro-2-methylbenzaldehyde SMILES: CC1=C(C=C(C=C1)F)C=O