missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (+)-5-Fluoro-2'-deoxyuridine, 99+%

CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol

Supplier:  Thermo Scientific Chemicals 227600025

Quantity 2.5 g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC227600025


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-91-9
246.19
ODKNJVUHOIMIIZ-RRKCRQDMSA-N
5790
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
C9H11FN2O5
MFCD00006530
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
CHEBI:60761
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

(+)-5-Fluoro-2′-deoxyuridine
148.0°C to 153.0°C
0.5% (60°C, 4 hrs) (vacuum) max.
99% min. (HPLC)
C9H11FN2O5
2.5 g
+ 35.90 (20.00°C c=1,water)
floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
ODKNJVUHOIMIIZ-RRKCRQDMSA-N
5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
5790
246.19
Powder
50-91-9
White
Authentic
Glass bottle
MFCD00006530
15,4144
+ 35.90
Solubility in water: soluble. Other solubilities: soluble in alcohol,practically insoluble in chcl3,insoluble in ether and hexane
OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O
246.19
CHEBI:60761
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
IF ON SKIN: Wash with plenty of soap and

GHS Signal Word: Danger

EINECSNumber : 200-072-5

SDS
Documents

Documents

Promotions

Promotions