accessibility menu, dialog, popup

UserName

CAS RN 50-91-9

IUPAC Name: 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: floxuridine|fluorodeoxyuridine|5 fluorodeoxyuridine|5-fluorodeoxyuridine|β-5-fluoro-2'-deoxyuridine|FdUrd|FdUrD|fdurd|FUdR|FUDR
Molecular Weight (g/mol): 246.19
Molecular Formula: C9H11FN2O5
InChi Key: ODKNJVUHOIMIIZ-RRKCRQDMSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (5)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)