missing translation for 'onlineSavingsMsg'
Learn More

5-(Ethylthio)-1H-tetrazole 98.0+%, TCI America™

Catalog No. E06701G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06701G 1 g
E06705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06701G Supplier TCI America Supplier No. E06701G

Chemical Identifiers

CAS 89797-68-2
Molecular Formula C3H6N4S
Molecular Weight (g/mol) 130.169
MDL Number MFCD00068733
InChI Key GONFBOIJNUKKST-UHFFFAOYSA-N
PubChem CID 2758928
IUPAC Name 5-ethylsulfanyl-2H-tetrazole
SMILES CCSC1=NNN=N1

Specifications

Melting Point 88°C
Quantity 1 g
UN Number 1325
Formula Weight 130.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Ethylthio)-1H-tetrazole