missing translation for 'onlineSavingsMsg'
Learn More

5-Cyano-2-methoxybenzaldehyde, 97%

Catalog No. AAH66690MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66690MD 250 mg
AAH6669003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66690MD Supplier Thermo Scientific Chemicals Supplier No. H66690MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21962-53-8 | C9H7NO2 | 161.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21962-53-8
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD13195275
InChI Key HJDXZGCTGAWUFZ-UHFFFAOYSA-N
Synonym 5-cyano-2-methoxybenzaldehyde, 3-formyl-4-methoxy-benzonitrile, 3-cyano6-methoxybenzaldehyde, 5-cyano-2-methoxy benzaldehyde
PubChem CID 10241098
IUPAC Name 3-formyl-4-methoxybenzonitrile
SMILES COC1=C(C=C(C=C1)C#N)C=O

Specifications

Melting Point 115°C to 120°C
Color Yellow
Quantity 250 mg
UN Number UN3439
Sensitivity Air Sensitive
Formula Weight 161.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Cyano-2-methoxybenzaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only