accessibility menu, dialog, popup

UserName

CAS RN 21962-53-8

IUPAC Name: 3-formyl-4-methoxybenzonitrile
Synonyms: 3-formyl-4-methoxybenzonitrile
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: HJDXZGCTGAWUFZ-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results