missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorouracil 98.0+%, TCI America™

Supplier:  TCI America C09695G

 View more versions of this product

Catalog No. C09695G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Chlorouracil
White-Yellow
MFCD00006019
5-chlorouracil, 2,4 1h,3h-pyrimidinedione, 5-chloro, uracil, 5-chloro, 5-chloro-2,4-dihydroxypyrimidine, 5-chloropyrimidine-2,4 1h,3h-dione, unii-7lq4v03rny, 7lq4v03rny, 5-chloro-1,3-dihydropyrimidine-2,4-dione, 5-chloropyrimidine-2,4 1h,3h-dione 5-chlorouracil, zlchem 428
ClC1=CNC(=O)NC1=O
146.53
CHEBI:60762
≥98.0% (T)
1820-81-1
C4H3ClN2O2
5 g
ZFTBZKVVGZNMJR-UHFFFAOYSA-N
5-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione
15758
146.53
Crystalline Powder

Chemical Identifiers

1820-81-1
146.53
ZFTBZKVVGZNMJR-UHFFFAOYSA-N
15758
5-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione
C4H3ClN2O2
MFCD00006019
5-chlorouracil, 2,4 1h,3h-pyrimidinedione, 5-chloro, uracil, 5-chloro, 5-chloro-2,4-dihydroxypyrimidine, 5-chloropyrimidine-2,4 1h,3h-dione, unii-7lq4v03rny, 7lq4v03rny, 5-chloro-1,3-dihydropyrimidine-2,4-dione, 5-chloropyrimidine-2,4 1h,3h-dione 5-chlorouracil, zlchem 428
CHEBI:60762
ClC1=CNC(=O)NC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : YQ9410000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions