accessibility menu, dialog, popup

UserName

ClNHOHNO

CAS RN 1820-81-1

IUPAC Name: 5-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-chlorouracil
Molecular Weight (g/mol): 146.53
Molecular Formula: C4H3ClN2O2
InChi Key: ZFTBZKVVGZNMJR-UHFFFAOYSA-N
SMILES: ClC1=CNC(=O)NC1=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)