missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorouracil 98.0+%, TCI America™

Catalog No. p-7124714
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
C09695G 5 g
C096925G 25 g
2 options

Catalog No. C09695G

Supplier: TCI America C09695G

Only null left

Chemical Identifiers

CAS 1820-81-1
Molecular Formula C4H3ClN2O2
Molecular Weight (g/mol) 146.53
MDL Number MFCD00006019
InChI Key ZFTBZKVVGZNMJR-UHFFFAOYSA-N
Synonym 5-chlorouracil, 2,4 1h,3h-pyrimidinedione, 5-chloro, uracil, 5-chloro, 5-chloro-2,4-dihydroxypyrimidine, 5-chloropyrimidine-2,4 1h,3h-dione, unii-7lq4v03rny, 7lq4v03rny, 5-chloro-1,3-dihydropyrimidine-2,4-dione, 5-chloropyrimidine-2,4 1h,3h-dione 5-chlorouracil, zlchem 428
PubChem CID 15758
ChEBI CHEBI:60762
IUPAC Name 5-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES ClC1=CNC(=O)NC1=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 146.53
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorouracil
RTECSNumber YQ9410000
TSCA No