missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chlorosaligenyl-N,N-diisopropylphosphoramidite 95.0+%, TCI America™
$249.78 - $721.40
Chemical Identifiers
| CAS | 1620086-77-2 |
|---|---|
| Molecular Formula | C13H19ClNO2P |
| Molecular Weight (g/mol) | 287.724 |
| InChI Key | ZJPWJYRCWIYHMH-UHFFFAOYSA-N |
| Synonym | 6-Chloro-N,N-diisopropyl-4H-benzo[d][1,3,2]dioxaphosphinin-2-amine |
| PubChem CID | 90306628 |
| IUPAC Name | 6-chloro-N,N-di(propan-2-yl)-4H-1,3,2-benzodioxaphosphinin-2-amine |
| SMILES | CC(C)N(C(C)C)P1OCC2=C(O1)C=CC(=C2)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C3326200MG
|
TCI America
C3326200MG |
200 mg |
Each for $249.78
|
|
|||||
|
C33261G
|
TCI America
C33261G |
1 g |
Each for $721.40
|
|
|||||
Chemical Identifiers
| 1620086-77-2 | |
| 287.724 | |
| 6-Chloro-N,N-diisopropyl-4H-benzo[d][1,3,2]dioxaphosphinin-2-amine | |
| 6-chloro-N,N-di(propan-2-yl)-4H-1,3,2-benzodioxaphosphinin-2-amine |
| C13H19ClNO2P | |
| ZJPWJYRCWIYHMH-UHFFFAOYSA-N | |
| 90306628 | |
| CC(C)N(C(C)C)P1OCC2=C(O1)C=CC(=C2)Cl |
Specifications
| 1620086-77-2 | |
| White | |
| 200 mg | |
| ZJPWJYRCWIYHMH-UHFFFAOYSA-N | |
| 6-chloro-N,N-di(propan-2-yl)-4H-1,3,2-benzodioxaphosphinin-2-amine | |
| 90306628 | |
| ≥95.0% (HPLC) | |
| 5-Chlorosaligenyl-N,N-diisopropylphosphoramidite |
| 45°C | |
| C13H19ClNO2P | |
| 6-Chloro-N,N-diisopropyl-4H-benzo[d][1,3,2]dioxaphosphinin-2-amine | |
| CC(C)N(C(C)C)P1OCC2=C(O1)C=CC(=C2)Cl | |
| 287.724 | |
| 287.72 | |
| Crystalline Powder |
Safety and Handling
TSCA : No