missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorosaligenyl-N,N-diisopropylphosphoramidite 95.0+%, TCI America™

Supplier:  TCI America C33261G

 View more versions of this product

Catalog No. C33261G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

5-Chlorosaligenyl-N,N-diisopropylphosphoramidite
45°C
C13H19ClNO2P
6-Chloro-N,N-diisopropyl-4H-benzo[d][1,3,2]dioxaphosphinin-2-amine
CC(C)N(C(C)C)P1OCC2=C(O1)C=CC(=C2)Cl
287.724
287.72
Crystalline Powder
1620086-77-2
White
1 g
ZJPWJYRCWIYHMH-UHFFFAOYSA-N
6-chloro-N,N-di(propan-2-yl)-4H-1,3,2-benzodioxaphosphinin-2-amine
90306628
≥95.0% (HPLC)

Chemical Identifiers

1620086-77-2
287.724
6-Chloro-N,N-diisopropyl-4H-benzo[d][1,3,2]dioxaphosphinin-2-amine
6-chloro-N,N-di(propan-2-yl)-4H-1,3,2-benzodioxaphosphinin-2-amine
C13H19ClNO2P
ZJPWJYRCWIYHMH-UHFFFAOYSA-N
90306628
CC(C)N(C(C)C)P1OCC2=C(O1)C=CC(=C2)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions