missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-6-hydroxynicotinic acid, 98%

CAS: 54127-63-8 | C6H3ClNO3 | 172.54 g/mol

$49.07 - $164.18

Chemical Identifiers

CAS 54127-63-8
Molecular Formula C6H3ClNO3
Molecular Weight (g/mol) 172.54
MDL Number MFCD00052950
InChI Key OLTRUTPHSBQWAZ-UHFFFAOYSA-M
Synonym 5-chloro-6-hydroxynicotinic acid, 3-chloro-2-hydroxypyridine-5-carboxylic acid, 5-chloro-6-hydroxypyridine-3-carboxylic acid, 5-chloro-6-hydroxy-3-pyridinecarboxylic acid, 5-chloro-6-hydroxynicotinicacid, 3-pyridinecarboxylic acid, 5-chloro-1,6-dihydro-6-oxo, 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid, pubchem10618, maybridge1_008981, acmc-1b1yw
PubChem CID 599541
IUPAC Name 5-chloro-6-oxo-1H-pyridine-3-carboxylic acid
SMILES [O-]C(=O)C1=CNC(=O)C(Cl)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1573706
View Documents
Thermo Scientific Chemicals
A1573706
5 g
Each for $49.07
Only null left
 
AAA1573714
View Documents
Thermo Scientific Chemicals
A1573714
25 g
Each for $164.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

54127-63-8
172.54
OLTRUTPHSBQWAZ-UHFFFAOYSA-M
599541
[O-]C(=O)C1=CNC(=O)C(Cl)=C1
C6H3ClNO3
MFCD00052950
5-chloro-6-hydroxynicotinic acid, 3-chloro-2-hydroxypyridine-5-carboxylic acid, 5-chloro-6-hydroxypyridine-3-carboxylic acid, 5-chloro-6-hydroxy-3-pyridinecarboxylic acid, 5-chloro-6-hydroxynicotinicacid, 3-pyridinecarboxylic acid, 5-chloro-1,6-dihydro-6-oxo, 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid, pubchem10618, maybridge1_008981, acmc-1b1yw
5-chloro-6-oxo-1H-pyridine-3-carboxylic acid

Specifications

54127-63-8
C6H3ClNO3
5 g
OLTRUTPHSBQWAZ-UHFFFAOYSA-M
5-chloro-6-oxo-1H-pyridine-3-carboxylic acid
599541
98%
>300°C
MFCD00052950
5-chloro-6-hydroxynicotinic acid, 3-chloro-2-hydroxypyridine-5-carboxylic acid, 5-chloro-6-hydroxypyridine-3-carboxylic acid, 5-chloro-6-hydroxy-3-pyridinecarboxylic acid, 5-chloro-6-hydroxynicotinicacid, 3-pyridinecarboxylic acid, 5-chloro-1,6-dihydro-6-oxo, 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid, pubchem10618, maybridge1_008981, acmc-1b1yw
[O-]C(=O)C1=CNC(=O)C(Cl)=C1
172.54
173.56
5-Chloro-6-hydroxynicotinic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only