accessibility menu, dialog, popup

UserName

CAS RN 54127-63-8

IUPAC Name: 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylate
Synonyms: 5-chloro-6-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 172.54
Molecular Formula: C6H3ClNO3
InChi Key: OLTRUTPHSBQWAZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CNC(=O)C(Cl)=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)