missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-6-hydroxynicotinic acid, 98%

CAS: 54127-63-8 | C6H3ClNO3 | 172.54 g/mol

Supplier:  Thermo Scientific Chemicals A1573714

 View more versions of this product

Catalog No. AAA1573714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chloro-6-hydroxynicotinic acid
>300°C
MFCD00052950
5-chloro-6-hydroxynicotinic acid, 3-chloro-2-hydroxypyridine-5-carboxylic acid, 5-chloro-6-hydroxypyridine-3-carboxylic acid, 5-chloro-6-hydroxy-3-pyridinecarboxylic acid, 5-chloro-6-hydroxynicotinicacid, 3-pyridinecarboxylic acid, 5-chloro-1,6-dihydro-6-oxo, 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid, pubchem10618, maybridge1_008981, acmc-1b1yw
[O-]C(=O)C1=CNC(=O)C(Cl)=C1
172.54
173.56
54127-63-8
C6H3ClNO3
25 g
OLTRUTPHSBQWAZ-UHFFFAOYSA-M
5-chloro-6-oxo-1H-pyridine-3-carboxylic acid
599541
98%

Chemical Identifiers

54127-63-8
172.54
OLTRUTPHSBQWAZ-UHFFFAOYSA-M
599541
[O-]C(=O)C1=CNC(=O)C(Cl)=C1
C6H3ClNO3
MFCD00052950
5-chloro-6-hydroxynicotinic acid, 3-chloro-2-hydroxypyridine-5-carboxylic acid, 5-chloro-6-hydroxypyridine-3-carboxylic acid, 5-chloro-6-hydroxy-3-pyridinecarboxylic acid, 5-chloro-6-hydroxynicotinicacid, 3-pyridinecarboxylic acid, 5-chloro-1,6-dihydro-6-oxo, 5-chloro-6-oxo-1,6-dihydropyridine-3-carboxylic acid, pubchem10618, maybridge1_008981, acmc-1b1yw
5-chloro-6-oxo-1H-pyridine-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only