missing translation for 'onlineSavingsMsg'
Learn More

5'-Chloro-3-hydroxy-2'-methyl-2-naphthanilide 96.0+%, TCI America™

Catalog No. C210225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C210225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C210225G Supplier TCI America Supplier No. C210225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135-63-7
Molecular Formula C18H14ClNO2
Molecular Weight (g/mol) 311.765
MDL Number MFCD00021636
InChI Key XZOACPDZZYNJER-UHFFFAOYSA-N
Synonym N-(5-Chloro-o-tolyl)-3-hydroxy-2-naphthamide, Naphthol AS-KB, Azoic Coupling Component 21, 5′C-Chloro-N-(3-hydroxy-2-naphthoyl)-o-toluidine
PubChem CID 67275
IUPAC Name N-(5-chloro-2-methylphenyl)-3-hydroxynaphthalene-2-carboxamide
SMILES CC1=C(C=C(C=C1)Cl)NC(=O)C2=CC3=CC=CC=C3C=C2O

Specifications

Melting Point 244°C
Quantity 25 g
Formula Weight 311.77
Percent Purity ≥96.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Chloro-3-hydroxy-2′-methyl-2-naphthanilide