CAS RN 135-63-7
IUPAC Name:
N-(5-chloro-2-methylphenyl)-3-hydroxynaphthalene-2-carboxamide
Synonyms:
N-(5-chloro-2-methylphenyl)-3-hydroxynaphthalene-2-carboxamide
Molecular Weight (g/mol):
311.77
Molecular Formula:
C18H14ClNO2
InChi Key:
XZOACPDZZYNJER-UHFFFAOYSA-N
SMILES:
CC1=CC=C(Cl)C=C1NC(=O)C1=C(O)C=C2C=CC=CC2=C1
1
–
1
of
1
results
1
5'-Chloro-3-hydroxy-2'-methyl-2-naphthanilide 96.0+%, TCI America™
CAS: 135-63-7 Molecular Formula: C18H14ClNO2 Molecular Weight (g/mol): 311.765 MDL Number: MFCD00021636 InChI Key: XZOACPDZZYNJER-UHFFFAOYSA-N Synonym: N-(5-Chloro-o-tolyl)-3-hydroxy-2-naphthamide, Naphthol AS-KB, Azoic Coupling Component 21, 5′C-Chloro-N-(3-hydroxy-2-naphthoyl)-o-toluidine PubChem CID: 67275 IUPAC Name: N-(5-chloro-2-methylphenyl)-3-hydroxynaphthalene-2-carboxamide SMILES: CC1=C(C=C(C=C1)Cl)NC(=O)C2=CC3=CC=CC=C3C=C2O