missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitrodiphenylamine 98.0+%, TCI America™

Catalog No. p-7125166
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17855G 5 g
C178525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C17855G Supplier TCI America Supplier No. C17855G

Chemical Identifiers

CAS 25781-92-4
Molecular Formula C12H9ClN2O2
Molecular Weight (g/mol) 248.666
MDL Number MFCD00007287
InChI Key FPKHZBVGKMTUHB-UHFFFAOYSA-N
PubChem CID 117635
IUPAC Name 5-chloro-2-nitro-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=C(C=CC(=C2)Cl)[N+](=O)[O-]

Specifications

Melting Point 114°C
Quantity 5 g
Synonym 5-chloro-2-nitrodiphenylamine, benzenamine, 5-chloro-2-nitro-n-phenyl, 5-chloro-2-nitrophenyl phenylamine, ghl.pd_mitscher_leg0.920, ksc203m3b, 2-anilino-4-chloronitrobenzene, fpkhzbvgkmtuhb-uhfffaoysa, 5-chloro-2-nitro-n-phenyl-benzenamine, benzenamine,5-chloro-2-nitro-n-phenyl
Formula Weight 248.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitrodiphenylamine