accessibility menu, dialog, popup

UserName

CAS RN 25781-92-4

ONOClHN

CAS RN 25781-92-4

IUPAC Name: 5-chloro-2-nitro-N-phenylaniline
Synonyms: 5-chloro-2-nitro-N-phenylaniline
Molecular Weight (g/mol): 248.67
Molecular Formula: C12H9ClN2O2
InChi Key: FPKHZBVGKMTUHB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C=C1NC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results