missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitrobenzamide 90.0+%, TCI America™

Catalog No. p-7125219
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C19785G 5 g
C197825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C19785G Supplier TCI America Supplier No. C19785G
Only null left

Chemical Identifiers

CAS 40763-96-0
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
InChI Key MKHXTOPPKVFSFI-UHFFFAOYSA-N
Synonym benzamide, 5-chloro-2-nitro, acmc-209jgl, 5-chloro-2-nitrobenzamid, 2-nitro-5-chlorobenzamide, 2-carbamoyl-4-chloronitrobenzene, 5-chloro-2-nitrobenzamide
PubChem CID 2723672
IUPAC Name 5-chloro-2-nitrobenzamide
SMILES C1=CC(=C(C=C1Cl)C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 154°C
Quantity 5 g
Formula Weight 200.58
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitrobenzamide