missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-nitrobenzamide, 98+%

Catalog No. AAL0938004
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0938004 2 g
AAL0938009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0938004 Supplier Thermo Scientific Chemicals Supplier No. L0938004
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 40763-96-0 | C7H5ClN2O3 | 200.578 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40763-96-0
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
MDL Number MFCD00017509
InChI Key MKHXTOPPKVFSFI-UHFFFAOYSA-N
Synonym benzamide, 5-chloro-2-nitro, acmc-209jgl, 5-chloro-2-nitrobenzamid, 2-nitro-5-chlorobenzamide, 2-carbamoyl-4-chloronitrobenzene, 5-chloro-2-nitrobenzamide
PubChem CID 2723672
IUPAC Name 5-chloro-2-nitrobenzamide
SMILES C1=CC(=C(C=C1Cl)C(=O)N)[N+](=O)[O-]

Specifications

Melting Point 155°C to 159°C
Quantity 2 g
Beilstein 2267797
Formula Weight 200.58
Percent Purity ≥98%
Chemical Name or Material 5-Chloro-2-nitrobenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only