missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™
$179.05 - $179.05
Chemical Identifiers
| CAS | 3438-16-2 |
|---|---|
| Molecular Formula | C8H7ClO3 |
| Molecular Weight (g/mol) | 186.591 |
| MDL Number | MFCD00017546 |
| InChI Key | HULDRQRKKXRXBI-UHFFFAOYSA-N |
| Synonym | 5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid |
| PubChem CID | 76970 |
| IUPAC Name | 5-chloro-2-methoxybenzoic acid |
| SMILES | COC1=C(C=C(C=C1)Cl)C(=O)O |
Chemical Identifiers
| 3438-16-2 | |
| 186.591 | |
| HULDRQRKKXRXBI-UHFFFAOYSA-N | |
| 76970 | |
| COC1=C(C=C(C=C1)Cl)C(=O)O |
| C8H7ClO3 | |
| MFCD00017546 | |
| 5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid | |
| 5-chloro-2-methoxybenzoic acid |
Specifications
| 3438-16-2 | |
| C8H7ClO3 | |
| 25 g | |
| HULDRQRKKXRXBI-UHFFFAOYSA-N | |
| 5-chloro-2-methoxybenzoic acid | |
| 76970 | |
| ≥98.0% (T) | |
| 5-Chloro-2-methoxybenzoic Acid |
| 99°C | |
| MFCD00017546 | |
| 5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid | |
| COC1=C(C=C(C=C1)Cl)C(=O)O | |
| 186.591 | |
| 186.59 | |
| Crystalline Powder |
Safety and Handling
TSCA : No