accessibility menu, dialog, popup

UserName

CAS RN 3438-16-2

H3COClOO

CAS RN 3438-16-2

IUPAC Name: 5-chloro-2-methoxybenzoate
Synonyms: 5-chloro-2-methoxybenzoate
Molecular Weight (g/mol): 185.58
Molecular Formula: C8H6ClO3
InChi Key: HULDRQRKKXRXBI-UHFFFAOYSA-M
SMILES: COC1=CC=C(Cl)C=C1C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results