missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. C153925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C153925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C153925G Supplier TCI America Supplier No. C153925G
Only null left

Chemical Identifiers

CAS 3438-16-2
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00017546
InChI Key HULDRQRKKXRXBI-UHFFFAOYSA-N
Synonym 5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid
PubChem CID 76970
IUPAC Name 5-chloro-2-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)Cl)C(=O)O

Specifications

Melting Point 99°C
Quantity 25 g
Formula Weight 186.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methoxybenzoic Acid