missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydroxybenzamide, 97%

CAS: 7120-43-6 | C7H6ClNO2 | 171.58 g/mol

Supplier:  Thermo Scientific Chemicals L1789706

 View more versions of this product

Catalog No. AAL1789706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

5-Chloro-2-hydroxybenzamide
223°C to 226°C
MFCD00007980
743348
NNHMQZBVJPQCAK-UHFFFAOYSA-N
5-chloro-2-hydroxybenzamide
348094
97%
7120-43-6
C7H6ClNO2
5 g
5-chlorosalicylamide, chlorosalicylamide, benzamide, 5-chloro-2-hydroxy, 5-chlorosalicyl-amide, acmc-209ohl, 5chloro-2-hydroxy-benzamide, ksc498a5j, 5-chloro-2-hydroxy-benzamide, #, 5-chloranyl-2-oxidanyl-benzamide
NC(=O)C1=CC(Cl)=CC=C1O
171.58
171.58

Chemical Identifiers

7120-43-6
171.58
NNHMQZBVJPQCAK-UHFFFAOYSA-N
348094
NC(=O)C1=CC(Cl)=CC=C1O
C7H6ClNO2
MFCD00007980
5-chlorosalicylamide, chlorosalicylamide, benzamide, 5-chloro-2-hydroxy, 5-chlorosalicyl-amide, acmc-209ohl, 5chloro-2-hydroxy-benzamide, ksc498a5j, 5-chloro-2-hydroxy-benzamide, #, 5-chloranyl-2-oxidanyl-benzamide
5-chloro-2-hydroxybenzamide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only