missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzylamine 97.0+%, TCI America™

Catalog No. C31585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31585G 5 g
C31581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31585G Supplier TCI America Supplier No. C31585G
Only null left

Chemical Identifiers

CAS 261723-26-6
Molecular Formula C7H7ClFN
Molecular Weight (g/mol) 159.59
MDL Number MFCD01631440
InChI Key BFLBLJKULFDOTN-UHFFFAOYSA-N
Synonym 5-chloro-2-fluorobenzylamine, 5-chloro-2-fluorophenyl methanamine, 2-fluoro-5-chlorobenzylamine, 2-fluoro-5-chlorobenzyl amine, 1-5-chloro-2-fluorophenyl methanamine, 5-chloro-2-fluoro-benzylamine, 5-chloro-2-fluorophenyl methylamine, 5-chloro-2-fluorobenzyl amine, pubchem4378
PubChem CID 2773616
IUPAC Name 1-(5-chloro-2-fluorophenyl)methanamine
SMILES NCC1=CC(Cl)=CC=C1F

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 159.59
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2-fluorobenzylamine