missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7125281
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20945G 5 g
C209425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20945G Supplier TCI America Supplier No. C20945G

Chemical Identifiers

CAS 394-30-9
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00665762
InChI Key WGAVMKXCDMQVNF-UHFFFAOYSA-N
Synonym 2-fluoro-5-chlorobenzoic acid, 5-chloro-2-fluoro-benzoic acid, 5-chloro-2-fluorobenzoicacid, benzoic acid, 5-chloro-2-fluoro, pubchem1375, acmc-1aigz, ksc223m7h, rarechem al bo 0944, 5-chloro-2-fluorobenzoic acid, benzoicacid, 5-chloro-2-fluoro
PubChem CID 2736537
IUPAC Name 5-chloro-2-fluorobenzoic acid
SMILES C1=CC(=C(C=C1Cl)C(=O)O)F

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-fluorobenzoic Acid