accessibility menu, dialog, popup

UserName

CAS RN 394-30-9

IUPAC Name: 5-chloro-2-fluorobenzoate
Synonyms: 5-chloro-2-fluorobenzoate
Molecular Weight (g/mol): 173.55
Molecular Formula: C7H3ClFO2
InChi Key: WGAVMKXCDMQVNF-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Cl)=CC=C1F
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)