missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%

CAS: 3380-34-5 | C12H7Cl3O2 | 289.536 g/mol

Supplier:  Thermo Scientific Chemicals L1865522

 View more versions of this product

Catalog No. AAL1865522


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
5-Chloro-2-(2,4-dichlorophenoxy)phenol, acts as a bacteriostat and preservative for cosmetic and detergent compositions. It is also used as an antiseptic, disinfectant and anticonvulsant. It is one of the main ingredient in shampoos, deodorants, toothpastes, mouth washes and household cleaning products. It inhibits the growth of bacteria, fungi on the surfaces of the textile, thereby it imparts odor protection to wool, synthetics, blends and non-wovens.

Solubility
Soluble in ethanol, methanol, diethyl ether and sodium hydroxide solution (1M). Slightly soluble in water.

Notes
Store in cool place. Decomposes on heating. Incompatible with strong oxidizing agents.
Specifications

Specifications

5-Chloro-2-(2,4-dichlorophenoxy)phenol
56°C to 59°C
MFCD00800992
UN3077
14,9657
Soluble in ethanol,methanol,diethyl ether and sodium hydroxide solution (1M). Slightly soluble in water.
C1=CC(=C(C=C1Cl)O)OC2=C(C=C(C=C2)Cl)Cl
289.536
CHEBI:164200
99%
3380-34-5
C12H7Cl3O2
100 g
2057142
triclosan, 5-chloro-2-2,4-dichlorophenoxy phenol, irgasan, 2,4,4'-trichloro-2'-hydroxydiphenyl ether, cloxifenolum, triclosanum, irgasan dp300, stri-dex face wash, aquasept, sapoderm
XEFQLINVKFYRCS-UHFFFAOYSA-N
5-chloro-2-(2,4-dichlorophenoxy)phenol
5564
289.55

Chemical Identifiers

3380-34-5
289.536
XEFQLINVKFYRCS-UHFFFAOYSA-N
5564
5-chloro-2-(2,4-dichlorophenoxy)phenol
C12H7Cl3O2
MFCD00800992
triclosan, 5-chloro-2-2,4-dichlorophenoxy phenol, irgasan, 2,4,4'-trichloro-2'-hydroxydiphenyl ether, cloxifenolum, triclosanum, irgasan dp300, stri-dex face wash, aquasept, sapoderm
CHEBI:164200
C1=CC(=C(C=C1Cl)O)OC2=C(C=C(C=C2)Cl)Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 222-182-2

RTECSNumber : KO1100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only