missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-(2,4-dichlorophenoxy)phenol, 99%

Catalog No. p-7047308
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1865509 10 g
AAL1865522 100 g
AAL1865536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL1865509 Supplier Thermo Scientific Chemicals Supplier No. L1865509
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 3380-34-5 | C12H7Cl3O2 | 289.536 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
5-Chloro-2-(2,4-dichlorophenoxy)phenol, acts as a bacteriostat and preservative for cosmetic and detergent compositions. It is also used as an antiseptic, disinfectant and anticonvulsant. It is one of the main ingredient in shampoos, deodorants, toothpastes, mouth washes and household cleaning products. It inhibits the growth of bacteria, fungi on the surfaces of the textile, thereby it imparts odor protection to wool, synthetics, blends and non-wovens.

Solubility
Soluble in ethanol, methanol, diethyl ether and sodium hydroxide solution (1M). Slightly soluble in water.

Notes
Store in cool place. Decomposes on heating. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 3380-34-5
Molecular Formula C12H7Cl3O2
Molecular Weight (g/mol) 289.536
MDL Number MFCD00800992
InChI Key XEFQLINVKFYRCS-UHFFFAOYSA-N
Synonym triclosan, 5-chloro-2-2,4-dichlorophenoxy phenol, irgasan, 2,4,4'-trichloro-2'-hydroxydiphenyl ether, cloxifenolum, triclosanum, irgasan dp300, stri-dex face wash, aquasept, sapoderm
PubChem CID 5564
ChEBI CHEBI:164200
IUPAC Name 5-chloro-2-(2,4-dichlorophenoxy)phenol
SMILES C1=CC(=C(C=C1Cl)O)OC2=C(C=C(C=C2)Cl)Cl

Specifications

Melting Point 56°C to 59°C
Quantity 10 g
UN Number UN3077
Beilstein 2057142
Merck Index 14,9657
Solubility Information Soluble in ethanol,methanol,diethyl ether and sodium hydroxide solution (1M). Slightly soluble in water.
Formula Weight 289.55
Percent Purity 99%
Chemical Name or Material 5-Chloro-2-(2,4-dichlorophenoxy)phenol
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 222-182-2
RTECSNumber KO1100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only