missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-1-pentanol 90.0+%, TCI America™
Supplier: TCI America C08155ML
Specifications
| 5-Chloro-1-pentanol | |
| Yellow | |
| C5H11ClO | |
| 5 mL | |
| DCBJCKDOZLTTDW-UHFFFAOYSA-N | |
| 5-chloropentan-1-ol | |
| 78915 | |
| ≥90.0% (GC) |
| 5259-98-3 | |
| 99°C | |
| MFCD00039556 | |
| 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l | |
| OCCCCCCl | |
| 122.59 | |
| 122.59 | |
| Liquid |
Chemical Identifiers
| 5259-98-3 | |
| 122.59 | |
| DCBJCKDOZLTTDW-UHFFFAOYSA-N | |
| 78915 | |
| OCCCCCCl |
| C5H11ClO | |
| MFCD00039556 | |
| 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l | |
| 5-chloropentan-1-ol |
Safety and Handling
EINECSNumber : (2)-2002
TSCA : Yes
Recommended Storage : Refrigerator