accessibility menu, dialog, popup

UserName

CAS RN 5259-98-3

IUPAC Name: 5-chloropentan-1-ol
Synonyms: 5-chloropentan-1-ol
Molecular Weight (g/mol): 122.59
Molecular Formula: C5H11ClO
InChi Key: DCBJCKDOZLTTDW-UHFFFAOYSA-N
SMILES: OCCCCCCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)