missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromovaleryl Chloride 98.0+%, TCI America™
$42.36 - $933.70
Chemical Identifiers
| CAS | 4509-90-4 |
|---|---|
| Molecular Formula | C14H11N3O2 |
| Molecular Weight (g/mol) | 253.26 |
| MDL Number | MFCD00013660 |
| InChI Key | QANQQXSQVQYQFI-UHFFFAOYSA-N |
| Synonym | 5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride |
| PubChem CID | 521173 |
| IUPAC Name | N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide |
| SMILES | CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1 |
Chemical Identifiers
| 4509-90-4 | |
| 253.26 | |
| QANQQXSQVQYQFI-UHFFFAOYSA-N | |
| 521173 | |
| CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1 |
| C14H11N3O2 | |
| MFCD00013660 | |
| 5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride | |
| N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide |
Specifications
| 4509-90-4 | |
| 118°C | |
| MFCD00013660 | |
| 3265 | |
| QANQQXSQVQYQFI-UHFFFAOYSA-N | |
| N-[2-(pyridin-4-yl)-1,3-benzoxazol-5-yl]acetamide | |
| 521173 | |
| ≥98.0% (T) | |
| 5-Bromovaleryl Chloride |
| Yellow | |
| C14H11N3O2 | |
| 5 g | |
| 5-bromovaleryl chloride, 5-bromovaleroyl chloride, 5-bromo-pentanoyl chloride, 5-bromovaleric acid chloride, 5-bromovalerylchloride, bromovalerylchloride, bromovaleryl chloride, 5-bromovaleric chloride, 5-bromopentanoylchloride, 5-bromo pentanoyl chloride | |
| CC(=O)NC1=CC=C2OC(=NC2=C1)C1=CC=NC=C1 | |
| 253.26 | |
| 199.47 | |
| Liquid |
Safety and Handling
TSCA : No